Learn More
BRAND™ Class B SILBERBRAND ETERNA Graduated Pipette
Class B SILBERBRAND ETERNA Graduated Pipette is constructed of AR-GLAS™ with amber graduations.
437.99 NOK - 650.00 NOK
Specifications
Aspirator Plug | Yes |
---|---|
Certifications/Compliance | DIN EN ISO 835 |
Class | B |
Description | To Deliver (TD, Ex.) |
Material | AR-GLAS |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15501526
|
BRAND™
27013 |
25 mL |
650.00 NOK
Pack of 6 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
15693947
|
BRAND™
27007 |
2 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- AR-GLAS™
- DIN EN ISO 835
- Calibrated to deliver (TD, Ex)
Specifications
Yes | |
B | |
AR-GLAS | |
Amber |
DIN EN ISO 835 | |
To Deliver (TD, Ex.) | |
Yes |