Learn More
Kartell™ Polypropylene Low Form Beakers with Molded Graduations
 Manufactured from Polypropylene for durability and good chemical resistance. Autoclavable Kartell™ Polypropylene Class B Low Form Beakers provide an economical and safe solution for stirring, mixing and dispensing liquids in the laboratory. 
Brand:  Kartell™ 0081100
Product Code. 11974299
  • 443.00 NOK / Pack of 2
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Description
  • Permanent molded or blue printed graduations provide clarity and reproducibility
  • Autoclavable at 121°C for 10 minutes
  • Withstands 100°C of continuous work
  • Class B, Low form
  • Complies with ISO 7056-1981 (E) and BS 5404 Part 1
  • Suitable for foodstuff
Product Title
Kartell™ Polypropylene Low Form Beakers with Molded Graduations > Capacity: 3000mL