Learn More
Honeywell™ Class 1 Plastic Cartridge A1B1E1 for Organic Vapors and Acid Gas
The K1 cartridge is part of the wide choice of cartridges offered to protect against many hazardous gases, vapors in many applications. The range provides high quality and a cost-effective protection.
Brand:  Honeywell™ N06575004L
Product Code. 15826635
  • 205.00 NOK / 1 pair
Estimated Shipment: 03-06-2025
Explore more special offers
Description
  • Protects against ammonia
  • Excellent filtration efficiency over extensive period of time
  • Lower profile and lightweight for optimal comfort
  • Low profile does not obstruct the field of vision
  • Threaded connector is easy to attach
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Class 1 plastic cartridge
Protects against ammonia
102.1 g
EC Category PPE: 3, QA: ISO 9001/2000, EC Certification: 2259, EC Attestation: 2259, EN 2259
6 Pair/Box, 8 Boxes/Cs.
Product Title
Honeywell™ Class 1 Plastic Cartridge A1B1E1 for Organic Vapors and Acid Gas > Protects against ammonia