Learn More
Honeywell™ Class 1 Plastic Cartridge A1B1E1 for Organic Vapors and Acid Gas
Click to view available options
:
Protects against organic vapors and acid gas
Description
- Protects against organic vapors and acid gas
- Excellent filtration efficiency over extensive period of time
- Lower profile and lightweight for optimal comfort
- Low profile does not obstruct the field of vision
- Threaded connector is easy to attach
Specifications
Specifications
Cartridge Type | Class 1 plastic cartridge |
Certifications/Compliance | EC Category PPE: 3, QA: ISO 9001/2000, EC Certification: 2259, EC Attestation: 2259, EN 2259 |
For Use With (Application) | Protects against organic vapors and acid gas |
Quantity | 6 Pair/Box, 8 Boxes/Cs. |
Weight (Metric) | 103.9 g |
Product Content Correction
Product Title
Honeywell™ Class 1 Plastic Cartridge A1B1E1 for Organic Vapors and Acid Gas >