missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIN85 Mouse anti-Human, Mouse, Rat, Clone: 3C12D2, Proteintech
Mouse Monoclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | CIN85 |
---|---|
Clone | 3C12D2 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16808575
|
Proteintech
66571-1-IG-20UL |
20 μL |
1611.84 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16898565
|
Proteintech
66571-1-IG-150UL |
150 μL |
4593.76 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes an adapter protein that contains three N-terminal Src homology domains, a proline rich region and a C-terminal coiled-coil domain. The encoded protein facilitates protein-protein interactions and has been implicated in numerous cellular processes including apoptosis, cytoskeletal rearrangement, cell adhesion and in the regulation of clathrin-dependent endocytosis. Alternate splicing results in multiple transcript variants.Specifications
CIN85 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CD2 binding protein 3, CD2BP3, CIN85, GIG10, HSB 1, MIG18, SH3KBP1 | |
Sh3kbp1 | |
IgG1 | |
Protein G | |
Antibody |
3C12D2 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
Q8R550, Q925Q9, Q96B97 | |
30011, 58194, 84357 | |
CIN85 Fusion Protein Ag2777 | |
Primary | |
-20°C | |
SH3KBP1 |