missing translation for 'onlineSavingsMsg'
Learn More
Chromium(Vi) Oxide, ReagentPlus™, Honeywell™ Product Code.: 15622310

Chromium(Vi) Oxide, ReagentPlus™, Honeywell™

Product Code. 15622310 Shop All Honeywell Research Chemicals Products
100 g, 100g
Click to view available options
Quantity:
5 g
100 g
1 kg
2.5 kg
Unit Size:
100g
1kg
2.50kg
5g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 15622310

Brand: Honeywell Research Chemicals 232653100G

Quantity Price
1 396.75 NOK / 100g
10 375.76 NOK / 100g

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.

View alternative products
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Reagent Grade, 99.9% trace metals basis

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1333-82-0
Molecular Formula CrO3
Molecular Weight (g/mol) 99.99
MDL Number MFCD00010952
InChI Key WGLPBDUCMAPZCE-UHFFFAOYSA-N
Synonym chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide
PubChem CID 14915
ChEBI CHEBI:48240
IUPAC Name trioxochromium
SMILES O=[Cr](=O)=O

Specifications

Quantity 100 g
UN Number 1463
Formula Weight 99.99g/mol
Percent Purity 99.9% (trace metals basis)
Grade ReagentPlus™
Chemical Name or Material Chromium(Vi) Oxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chromium(Vi) Oxide, ReagentPlus™, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.