Learn More
Macherey-Nagel™ Chromafix™ SA Cartridge
2105.00 NOK - 3265.00 NOK
Specifications
Base Material | Silica |
---|---|
For Use With (Application) | Amino Acids, Amines, Chlorophyll, PCBs |
Particle Size | 45 μm |
Sorbent | SA |
Type | Chromafix SA Cartridge |
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
10135091
|
Macherey-Nagel™
731832 |
Adsorbent Weight: 450 mg |
2450.00 NOK
Pack of 50 |
In Stock
Log in to see stock. |
||||||
10175041
|
Macherey-Nagel™
731831 |
Adsorbent Weight: 220 mg |
2105.00 NOK
Pack of 50 |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
10145091
|
Macherey-Nagel™
731833 |
Adsorbent Weight: 920 mg |
3265.00 NOK
Pack of 50 |
In Stock
Log in to see stock. |
||||||
Specifications
Silica | |
45 μm | |
Chromafix SA Cartridge | |
50 Pack |
Amino Acids, Amines, Chlorophyll, PCBs | |
SA | |
60 Å | |
500 m2/g |
Product Title
Macherey-Nagel™ Chromafix™ SA Cartridge