Learn More
Thermo Scientific™ Chromacol EPA Pre-Assembled Vials
Thermo Scientific™ Chromacol EPA Preassembled Vials include caps and seals.
Brand:  Thermo Scientific™ 20-EPAVCS-PC
2380.07 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10287962
  • 2395.00 NOK / Pack of 72
Only null left
Estimated Shipment: 13-08-2025
Description
Thermo Scientific™ Chromacol EPA Preassembled Vials include caps and seals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20mL Clear Glass EPA Vial with Cap and Seal Class 200 Pre-cleaned
Clear
20 mL
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Chromacol EPA Pre-Assembled Vials >