Learn More
Macherey-Nagel™ Chromabond™ Polyethylene Frit
Assemble SPE (solid-phase extraction) columns selecting from a range of accessories. Chromabond™ Polyethylene Frits for polyethelyne columns come in a varitey of sizes.
Brand:  Macherey-Nagel™ 730162
 View more versions of this product
Product Code. 10639581
  • 2165.00 NOK / Pack of 250
Only null left
Estimated Shipment: 04-07-2025
Description
Accessory for SPE (solid-phase extraction)
For individual packing of SPE columns with Chromabond™ adsorbents
Specifications
Specifications
Polypropylene Columns
3 mL
250 Pack
Frit
Polyethylene
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Macherey-Nagel™ Chromabond™ Polyethylene Frit > Volume: 3mL