Learn More
Macherey-Nagel™ Chromabond™ Polyethylene Frit
Assemble SPE (solid-phase extraction) columns selecting from a range of accessories. Chromabond™ Polyethylene Frits for polyethelyne columns come in a varitey of sizes.
Brand:  Macherey-Nagel™ 730163
 View more versions of this product
Product Code. 10154801
  • 2655.00 NOK / Pack of 250
Only null left
Estimated Shipment: 04-07-2025
Description
Accessory for SPE (solid-phase extraction)
For individual packing of SPE columns with Chromabond™ adsorbents
Specifications
Specifications
Polypropylene Columns
6 mL
250 Pack
Frit
Polyethylene
Documents
One moment while we fetch your results.
Product Certifications
Promotions
Promotions
Product Content Correction
Product Title
Macherey-Nagel™ Chromabond™ Polyethylene Frit > Volume: 6mL