Learn More
Chloroformates Gas Detection Tube
Brand:  Draeger Safety 6718601
Product Code. 10532804
  • 5945.00 NOK / Pack of 10
Explore more special offers
Specifications
5 to 15 mg H20/L
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Chloroformates 0.2/b
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.