missing translation for 'onlineSavingsMsg'
Learn More
Chloroform, MP Biomedicals™ Product Code.: 11428262

Chloroform, MP Biomedicals™

Product Code. 11428262 Shop All MP Biomedicals Products
4 L
Click to view available options
Quantity:
4 L
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11428262

Brand: MP Biomedicals 0219381404

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Chloroform is typically used as a solvent and as a cleansing agent.

Procedures have been described for the use of chloroform in lambda plaques storage, lambda cDNA storage, the removal of mineral oil from PCR reaction samples, oligonucleotide purification, a hydroxyl radical footprinting protocol, a transcriptional run-on assay protocol, and an overlay assay for beta-galactosidase activity.

TRUSTED_SUSTAINABILITY

Specifications

CAS 67-66-3
Quantity 4 L
Physical Form Liquid
Assay Percent Range ≥99.8%
Molecular Formula CHCl3
MDL Number MFCD00000826
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
Solubility Information Soluble in water (1mL/200mL water); miscible with ethanol, benzene, ether, petroleum ether, carbon tetrachloride, carbon disulfide, oils.
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
SMILES ClC(Cl)Cl
IUPAC Name trichloromethane
Molecular Weight (g/mol) 119.37
PubChem CID 6212
ChEBI CHEBI:35255
Formula Weight 119.4g/mol
Grade ACS
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chloroform, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.