missing translation for 'onlineSavingsMsg'
Learn More
Chloroform, Amylene Stabilized, HPLC Grade, Scharlab Product Code.: 11318374
compliance-icons compliance-icons compliance-icons

Chloroform, Amylene Stabilized, HPLC Grade, Scharlab

Product Code. 11318374
1 L, Glass Bottle, 1 L
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11318374

Brand: Scharlab CL02071000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Stabilized with approximately 150 ppm Amylene. Chloroform, Amylene Stabilized, HPLC Grade, Scharlab is of appropriate purity to be used as a mobile phase in HPLC in analytical and preparative separations.

  • Undergoes highly efficient distillation, solid phase absorption and other treatments to eliminate impurities that could interfere in HPLC analysis
  • Microfiltered and packed in amber-coloured bottles (sealed with a PTFE membrane in the cap) to avoid possible contamination

Specifications

Melting Point -63°C
Absorbance 0.699 AU at 248nm, 0.301 AU at 253nm, 0.046 AU at 265nm
CAS 67-66-3
Color Colorless
Acidity Max. 0.0002meq/g
Alkalinity Max. 0.0002meq/g
Density 1.487 - 1.490g/cm3
Boiling Point 61°C
Quantity 1 L
Identification Passes test
Ignition Point 982°C
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation Max. 0.0003%
Molecular Formula CHCl3
MDL Number MFCD00000826
Vapor Pressure 213 hPa
UN Number 1888
Water Max. 0.01%
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
Solubility Information 8g/L in water
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
SMILES ClC(Cl)Cl
IUPAC Name trichloromethane
Molecular Weight (g/mol) 119.37
PubChem CID 6212
ChEBI CHEBI:35255
Formula Weight 119.38g/mol
Percent Purity Min. 99.8%
Grade Applications: solvents, analytical chemistry, in the rubber industry
Chemical Name or Material Chloroform
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chloroform, HPLC grade, stabilized with amylene (approx. 150 ppm)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H331-Toxic if inhaled.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chloroform, Amylene Stabilized, HPLC Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.