Learn More
Thermo Scientific Chemicals Chemotactic Peptide
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Induces leucocyte chemotaxis, and activates macrophages
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Purification Method | HPLC |
Common Name | fMLP |
Content And Storage | -30°C to -10°C |
Format | Lyophilized Powder |
For Use With (Application) | Induces leucocyte chemotaxis, and activates macrophages |
Molecular Weight (g/mol) | 437.55 |
Product Type | Chemotactic Peptide |
Quantity | 25 mg |
Regulatory Status | RUO - research use only |
Sequence | N-Formyl-Met-Leu-Phe |
Product Title
Thermo Scientific Chemicals Chemotactic Peptide >