missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals Channel Catfish Diet, Powder
Powdered fish diet for channel catfish. Water must be added. Agar, casein, dextrin and salt mixture.
Brand: MP Biomedicals 0296027650
This item is not returnable.
View return policy
Description
- Powdered fish diet for channel catfish
- Water must be added
- Agar, casein, dextrin and salt mixture
- Contains MP Bio vitamin mixture
Specifications
Not Autoclavable | |
Single-use | |
50 kg | |
Room Temperature | |
0.3048 m |
Animal Diet | |
Powder | |
Non-sterile | |
1 ft. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction