Learn More
Nabertherm™ Chamber Furnace LF 1400 Series with B500 Controller
Combined insulation of refractory bricks in the corners and low heat storage and quickly cooling fiber material. With a wide variety of optional equipment, these chamber furnaces can be optimally adapted to your processes.
Brand:  Nabertherm™ LF0642NKNE
 View more versions of this product
Product Code. 17307949
  • 194865.00 NOK / Each
Only null left
Estimated Shipment: 11-08-2025
Description
  • Maximum Temperature: 1400°C
  • Dual shell housing with rear ventilation, provides for low shell temperatures
  • Five-sided heating for very good temperature uniformity
  • Heating elements on support tubes ensure free heat radiation and a long service life
  • Controller mounted on furnace door and removable for comfortable operation
  • Protection of bottom heating and flat stacking surface provided by embedded SiC plate in the floor
  • High-quality non-classified fiber insulation with corner bricks for shorter heating and cooling times
  • Door with brick-on-brick seal, hand fitted
  • Short heating times due to high installed power
  • Side vent with bypass connection for exhaust pipe
  • Self-supporting arch for high stability and greatest possible protection against dust
  • Quick lock on door
  • Freely adjustable air slide intake in furnace floor
  • Stand included
  • Defined application within the constraints of the operating instructions
Specifications
Specifications
B500
1400°C
1070 x 780 x 1365 mm
60 L
400 x 400 x 400 mm
300 kg
Documents
Brochures
Product Guides
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Nabertherm™ Chamber Furnace LF 1400 Series with B500 Controller > Capacity: 60 L