Learn More
Scharlab Cetrimide Agar (Eur. Pharm.)
Scharlab Cetrimide Agar (Eur. Pharm.) is used for selective isolation of Pseudomonas aeruginosa.
Brand:  Scharlab 01-160-500
Product Code. 11481767
  • 1915.00 NOK / 500g
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • AOAC/EP/ISO/HP/USP
  • Solid culture medium for selective isolation of Pseudomonas aeruginosa
  • According to Harmonised Pharmacopoeia Method and 22717 standard.
  • This medium needs glycerol (Art. No.: GL00261000)
Specifications
Specifications
22717 Standard
Cetrimide Agar
Gram-negative Bacteria
Selective Isolation of Pseudomonas Aeruginosa
18 to 72 hr.
500 g
Solid
Selective Medium
Gelatin Peptone 20.00 g/L, Magnesium Chloride 1.40 g/L, Potassium Sulphate 10.00 g/L, Cetyl-N,N,N-Trimethylammoniumbromide 0.30 g/L, Agar 13.60 g/L
30°C to 35°C
7.2 ±0.2
Insoluble