missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP70 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | CEP70 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16895864
|
Proteintech
16280-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16885864
|
Proteintech
16280-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CEP70 plays a role in the organization of both preexisting and nascent microtubules in interphase cells. During mitosis, it is required for the organization and orientation of the mitotic spindle.Specifications
CEP70 | |
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q5PQQ9, Q6IQY5, Q8NHQ1 | |
367153, 68121, 80321 | |
CEP70 Fusion Protein Ag9365 | |
Primary | |
-20°C | |
CEP70 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BITE, centrosomal protein 70kDa, Centrosomal protein of 70 kDa, CEP70, p10 binding protein | |
Cep70 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |