Learn More
Eppendorf™ Centrifuge Bucket
Designed for use with Eppendorf Rotors and Centrifuges
Brand: Eppendorf™ 5810716004
13100.00 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Buckets for: Rotor S-24-11-AT, Rotor A-4-62, Rotor A-4-81
Specifications
Eppendorf rotor A-4-62 (CFA-150-010C) | |
Bucket | |
4/Pk. |
250 mL | |
1 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Product Content Correction
Product Title
Eppendorf™ Centrifuge Bucket > For use with: rotor A-4-62 (CFA-150-010C); Quantity: 4/Pk.; Volume: 250mL