Learn More
Eppendorf™ Centrifuge Adapters
Designed for use with Eppendorf Rotors and Centrifuges
Brand: Eppendorf™ 5702702005
1010.00 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Accessory for Eppendorf liquid handling equipment
Specifications
Tube Adapter | |
Rotor A-8-17 | |
1 Conical Tube |
1 x 15 mL Conical Tube | |
Adapter | |
8/Pk. |
Product Content Correction
Product Title
Eppendorf™ Centrifuge Adapters > For Use With: Rotor A-8-17; Type: Adapter