Learn More
Greiner Bio-One CELLSTAR 384-well, Cell Culture-Treated, Flat-Bottom Microplate
Click to view available options
Packaging:
8/Pk., 32/Cs.
Description
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic Non-cytotoxicCompatible with automated systemsFlexible working volumeRounded square well profile
- Stackable
- Alphanumeric well cod
Specifications
Specifications
Format | Standard |
Lid | With Lid |
Array | 16 x 24 |
Packaging | 8/Pk., 32/Cs. |
Color | White |
Product Type | Solid Bottom |
Material | Polystyrene |
No. of Wells | 384 |
Volume (Metric) Working | 15 to 110 μL |
Sterility | Sterile |
Show More |
Product Content Correction
Product Title
Greiner Bio-One CELLSTAR 384-well, Cell Culture-Treated, Flat-Bottom Microplate >