Learn More
BINDER™ CELLROLL Set
CELLROLL Set is a modular, expandable roller bottle system for cell cultivation.
Brand:  BINDER™ 8612-0023
 View more versions of this product
Product Code. 17313683
  • 94485.00 NOK / Each
Estimated Shipment: 11-08-2025
Description
  • Adjustable direction of rotation, speed of rotation, pause times, and interval cycles
  • All drive rollers for the bottles run without vibrations on special ball bearings
  • Driven by a stepper motor with a soft start and stop phase
  • Control unit is outside the incubator and is connected to the interior motor unit via an electric access port
Specifications
Specifications
CELLROLL Set - modular, expandable roller bottle system for cell cultivation, 4 roller bottles
Stainless Steel
Cell Cultivation
4 Roller Bottles/Pk.
BINDER Series CB and CBF
Cell Roller
CBF 170
Documents
One moment while we fetch your results.
Certificates
View accessory products
One moment while we fetch your results.
Promotions
Promotions
Product Content Correction
Product Title
BINDER™ CELLROLL Set > Model: CBF 170, Quantity: 4 Roller Bottles/Pk.