Learn More
Molecular Biology Grade Water Tested to USP Sterile water, Corning™
Description
Molecular biology grade water is ideal for applications where highly pure, RNase-, DNase-, and protease-free water is required (e.g., PCR and other molecular biology reactions setup, DNA/RNA/protein extraction, purification, and storage reagents). Molecular biology grade water is tested to the chemical and physical properties found in the USP monograph for sterile purified water.
Certifications
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility
Specifications
Specifications
Quantity | 6 x 100 mL |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
32524006 | Certificate of Quality | 16/04/2025 | 46-000-CI |
29224008 | Certificate of Quality | 16/04/2025 | 46-000-CI |
26124011 | Certificate of Quality | 16/04/2025 | 46-000-CI |
22724008 | Certificate of Quality | 26/12/2024 | 46-000-CI |
13424007 | Certificate of Quality | 01/11/2024 | 46-000-CI |
Special Offers
-
Corning Matrigel
,
Corning Biocoat
,
Corning CoolProduct
,
Corning Media Tech
,
Corning
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70% -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30