Learn More
STEMCELL Technologies CellAdhere™ Laminin-521
Matrix for maintenance of human ES and iPS cells in combination with TeSR™ maintenance media
Brand:  STEMCELL Technologies 200-0117
Encompass_Preferred
Product Code. 17178001
  • 5762.37 NOK / 500µg
Description
  • Support the growth and differentiation of human embryonic stem (ES) and induced pluripotent stem (iPS) cells under feeder-free conditions with this defined and xeno-free cell culture matrix
  • For consistent cell populations and reproducible results in downstream applications, use CellAdhere™ Laminin-521 with TeSR™ maintenance media to provide a defined culture substrate for cell maintenance
  • Laminin 521 is expressed and secreted by human pluripotent stem cells (hPSCs) in the inner cell mass of the embryo and therefore creates a biologically relevant hPSC culture environment in vitro
  • Compared to other matrices, CellAdhere™ Laminin-521 increases single-cell attachment and survival and does not require the addition of apoptotic inhibitors during plating
  • Note that single-cell passaging of human ES and iPS cells can result in selective pressure and lead to genetic aberrations
  • If passaging as single cells, we recommend checking the karyotype frequently
Specifications
Specifications
Pluripotent Stem Cells
500 μg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies CellAdhere™ Laminin-521