Learn More
Fisherbrand™ Cell Holder for Eco 8/Eco 25 Thermoreactors
Helps facilitate the COD analysis process. Fisherbrand™ Cell Holders for Eco 8/Eco 25 Thermoreactors have the capacity for 12 round cells.
Brand: Fisherbrand™ CA0091636
456.25 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Securely holds glass test tubes
- Sample processing
Specifications
ECO 8/ECO 25 Thermoreactor | |
12 Tubes | |
1 Each | |
Holder, Cell |
Promotions
-
Multiple Manufacturers
Focus On Clean Water
Shop now for up to 60% off Clean Water Research essentialsPromo Code: 24139Expires: 2025-08-21
New Offer Save Now