Learn More
Hellma™ Cell
Brand:  Hellma™ 1767005-15-40
Product Code. 13297579
  • 10385.00 NOK / Each
Only null left
Estimated Shipment: 31-07-2025
Specifications
Cell
195 ÎĽL
19 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hellma™ Cell > Diameter: 19mm