Learn More
Vileda Professional™ CE MicroIntensive Duo™
Double-sided, single-use mop to speed-up the cleaning process, while also reducing the risk of cross-contamination.
5200.00 NOK - 5580.00 NOK
Specifications
Color White
Autoclavable Autoclavable
Material Polyester Microfiber
View More Specs
Products 2
Product Code Brand Sterility Price Quantity & Availability  
Product Code Brand Sterility Price Quantity & Availability  
17812476
View Documents
Vileda Professional™
166813
Non-sterile
5200.00 NOK
Pack of 25
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
17822476
View Documents
Vileda Professional™
166815
Sterile
5580.00 NOK
Pack of 25
Estimated Shipment: 22-01-2025
Description
  • Independent lab study, conducted by BMA Labs, resulting in bacteria reduction of 99.99%
  • 4-layer construction - featuring Polyester/Microfiber embossed cleaning side
  • Pre-laundered for single-use
  • Gamma and autoclave compatible
  • Extremely low particle shedding
  • Double-sided cleaning with color -stitched side indicator
  • Corner drainage openings for cleaner rinses
  • Finger loop for touchless on/off of cleaning sides
Specifications
Specifications
White
Autoclavable
Polyester Microfiber