missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC42BPB, Mouse anti-Human, Clone: 5F12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009578-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: SKMISNPTNFNHVAHMGPGDGMQVLMDLPLSAVPPSQEERPGPAPTNLARQPPSRNKPYISWPSSGGSEPSVTVPLRSMSDPDQDFDKEPDSDSTKHSTPSpecifications
CDC42BPB | |
Monoclonal | |
Unconjugated | |
CDC42 binding protein kinase beta (DMPK-like) | |
KIAA1124/MRCKB | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody |
ELISA, Immunofluorescence, Western Blot | |
5F12 | |
In 1x PBS, pH 7.4 | |
AF128625 | |
CDC42BPB | |
CDC42BPB (AAD37506, 1580 a.a. ∼ 1679 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
9578 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CDC42BPB, Mouse anti-Human, Clone: 5F12, Abnova™ > 100μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction