missing translation for 'onlineSavingsMsg'
Learn More

CD90 (Thy-1) Mouse, anti-Human, APC, Clone: 5E10, STEMCELL Technologies

Mouse Monoclonal Antibody

3239.77 NOK - 6890.43 NOK

Specifications

Antigen CD90 (Thy-1)
Clone 5E10
Applications Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Conjugate APC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17118761
Encompass_Preferred
View Documents
Stemcell Technologies
60045AZ.1
25 Tests
3239.77 NOK
25 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17108761
Encompass_Preferred
View Documents
Stemcell Technologies
60045AZ
100 Tests
6890.43 NOK
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • The 5E10 antibody reacts with CD90 (Thy-1), a GPI-linked membrane glycoprotein that is N-glycosylated at two sites, giving rise to 25 - 37 kDa molecules
  • CD90 has roles in signal transduction, cell adhesion and migration, neurite outgrowth, T cell activation, tumor suppression, and inhibition of the proliferation and differentiation of hematopoietic stem cells
  • It is a known ligand of β2 and β3 integrins and upregulates synthesis of fibronectin, osteonectin and thrombospondin
  • CD90 is broadly expressed, being found on human thymocytes, neurons, some glial cells, fibroblasts, activated endothelial cells, some leukemia cell lines and a distinct subset (<1%) of CD3+CD4+ T cells in human peripheral blood
  • This antibody clone has been verified for labeling human mesenchymal cells grown in MesenCult™ Proliferation Kit (Human; Catalog #05411) and MesenCult™-XF Medium (Catalog #05420)
Specifications

Specifications

CD90 (Thy-1)
Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry
APC
Baboon, Cynomolgus, Rhesus Monkey, Pig, Human
Primary
5E10
Monoclonal
Mouse
IgG1 κ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.