Learn More
CD85d (ILT4) Monoclonal Antibody (42D1), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
30070.00 NOK - 150400.00 NOK
Specifications
Antigen | CD85d (ILT4) |
---|---|
Clone | 42D1 |
Concentration | 1 mg/mL |
Applications | Flow Cytometry, Functional Assay |
Classification | Monoclonal |
Description
The MAb 42D1 recognizes the CD85d protein also known as ILT4, LIR-2. It is a member of the ILT (immunoglobulin-like transcript)/ LIR (leukocyte Ig-like receptor), MIR (monocytes Ig-like receptor) family. CD85d is a 110 kDa single transmembrane receptor with 3 Ig-like domains and a long cytoplasmic domain containing ITIMs (immunoreceptor tyrosine-based inhibitory motifs). Expression is confined to monocytes, macrophages and dendritic cells. The ligand for CD85d is non-classical class I HLA-G, -B and G1. Upon ligand binding, tyrosine phosphatase SHP-1 interacts with the ITIM. Interaction of HLA-G with ILT4/CD85d is thought to contribute to tolerance during fetal implantation. Additionally, it is the levels of ILT3 and ILT4 on DCs that suggests a role in organ allograft rejection. Crosslinking of the receptor with the MAb 42D1 inhibits calcium flux as well as enhancing HLA-G tetramer staining and restoring the stimulating activity of tolerogenic DC.
CD85d (ILT4) Monoclonal antibody specifically detects CD85d (ILT4) in Human samples. It is validated for Flow Cytometry, Functional AssaySpecifications
CD85d (ILT4) | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.2 | |
CD85 antigen-like family member D; CD85D; CD85d antigen; Ig-like transcript 4; ILT4; ILT-4; leucocyte Ig-like receptor B2; leukocyte immunoglobulin like receptor B2; leukocyte immunoglobulin-like receptor subfamily B member 2; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 2; LILRB2; LIR2; LIR-2; LIR2CD85D; MIR10; MIR-10; MIR10LILRA6; monocyte/macrophage immunoglobulin-like receptor 10; myeloid inhibitory receptor 10 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
42D1 | |
Flow Cytometry, Functional Assay | |
Functional Grade | |
Rat | |
Human | |
Q8N423 | |
10288 | |
Primary | |
4° C | |
LILRB2 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.