missing translation for 'onlineSavingsMsg'
Learn More

CD80 (B7-1) Monoclonal Antibody (2D10.4), Super Bright™ 702, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

1712.00 NOK - 3220.00 NOK

Specifications

Antigen CD80 (B7-1)
Clone 2D10.4
Concentration 5 μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16391634
View Documents
eBioscience
67-0809-41
25 Tests
1712.00 NOK
Pack of 25
Please to purchase this item. Need a web account? Register with us today!
16301644
View Documents
eBioscience
67-0809-42
100 Tests
3220.00 NOK
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: The 2D10.4 monoclonal antibody reacts with human CD80 (B7-1), a 60 kDa member of the Ig superfamily. CD80 is expressed by activated B cells, macrophages and dendritic cells. In addition, activated T cells express this antigen. CD80 has high affinity for binding to two T cell surface antigens, CD28 and CD152 (CTLA-4). The interaction of CD28 and CD152 with CD80 is crucial in T-B cell communication leading to activation of T and B cells, respectively. Applications Reported: This 2D10.4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 2D10.4 antibody has been pre-diluted and tested by flow cytometric analysis of Daudi cell line. This may be used at 5 μL (1.0 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 702 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 702 nm. We recommend using a 710/50 bandpass filter.  Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product No. SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information.

CD80 (B7-1) and CD86 (B7-2) are ligands of T cell critical costimulatory molecule CD28, and of an inhibitory receptor CTLA-4 (CD152). Both B7 molecules are expressed on professional antigen-presenting cells and are essential for T cell activation, and both molecules can also substitute for each other in this process. CD80 is rapidly induced on the surface of in vitro activated B cells, with Epstein Barr Virus (EBV) transformed B cell lines, with Burkitts lymphoma cell lines, with freshly isolated follicular B lymphoma cells, T cells, and monocytes. The B-lymphocyte activation antigen B7-1 provides regulatory signals for T lymphocytes as a consequence of binding to the CD28 and CTLA4 ligands of T cells. Diseases associated with CD80 dysfunction include gallbladder squamous cell carcinoma and myocarditis.
Specifications

Specifications

CD80 (B7-1)
5 μL/Test
Monoclonal
Liquid
RUO
P33681
CD80
Primary
Antibody
2D10.4
Flow Cytometry
Super Bright 702
Mouse
Human
941
IgG1 κ
4° C, store in dark, DO NOT FREEZE!
CD80
Videos
SDS
Documents

Documents

Product Certifications