Learn More
CD62L (L-Selectin) Monoclonal Antibody (MEL-14), Brilliant Ultra Violet™ 615, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
1716.00 NOK - 3515.00 NOK
Specifications
Antigen | CD62L (L-Selectin) |
---|---|
Clone | MEL-14 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17847050
|
Invitrogen
366-0621-80 |
25 μg |
1716.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17857050
|
Invitrogen
366-0621-82 |
100 μg |
3515.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MEL-14 mAb reacts with mouse CD62L, a 76 kDa member of the selectin family. CD62L is expressed by neutrophils, monocytes, and subsets of T, B, and NK cells and binds a number of glycosylated, fucosylated, sulfated sialylated glycoproteins including CD34, glycam-1 and MAdCam-1. These interactions mediate rolling of lymphocytes on activated endothelium at the sites of inflammation and homing of cells to the high endothelial venules (HEV) of peripheral lymphoid tissues. Applications Reported: This MEL-14 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MEL-14 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.06μg per test Brilliant Ultra Violet 615 (BUV615) is a tandem dye that emits at 615 nm and is intended for use on cytometers equipped with an Ultraviolet (355 nm) laser. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright, Brilliant Violet, Brilliant Ultra Violet, or other polymer dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (SB-4401-42) or Brilliant Stain Buffer (00-4409-75) to minimize any non-specific polymer interactions. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Excitation: 350 nm; Emission: 615 nm; Laser: Ultraviolet Laser.
The human CD62L is a 74-95 kDa glycoprotein member of the selectin family of adhesion receptors. L-Selectin is comprised of an aminoterminal C-type lectin binding domain, an epidermal growth factor-like domain, two short consensus repeat (SCR) sequences homologous to those found in complement binding proteins, a short spacer region, a transmembrane region and a short cytoplasmic region. Human CD62L (L-Selectin) is constitutively expressed on all classes of leukocytes including lymphocytes (except a substantial population of memory T cells), monocytes and polymorphonuclear cells.Specifications
CD62L (L-Selectin) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
A.11; AI528707; CD62 antigen-like family member L; CD62L; gp90-MEL; hLHRc; LAM1; LAM-1; LECAM1; LECAM-1; LEU8; Leu-8; Leukocyte adhesion molecule 1; leukocyte surface antigen Leu-8; Leukocyte-endothelial cell adhesion molecule 1; LNHR; LSEL; L-selectin; Ly22; Ly-22; LYAM1; Lyam-1; Ly-m22; Lymph node homing receptor; lymphocyte adhesion molecule 1; lymphocyte antigen 22; Lymphocyte surface MEL-14 antigen; pln homing receptor; PLNHR; SELE; selectin E (endothelial adhesion molecule 1); selectin L; selectin L (lymphocyte adhesion molecule 1); selectin, lymphocyte; Sell; TQ1 | |
SELL | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
SELL |
MEL-14 | |
Flow Cytometry | |
Brilliant Ultraviolet 615 | |
Rat | |
Mouse | |
P18337 | |
20343 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |