missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD59 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10742-1-AP-150UL
This item is not returnable.
View return policy
Description
CD59 (Protectin) is a small (18-20 kDa) GPI-anchored ubiquitously expressed inhibitor of the membrane attack complex (MAC). CD59 is the key regulator that preserves the autologous cells from terminal effector mechanism of the complement cascade. CD59 also associates with C5b-8 complex and counteracts appropriate formation of cytolytic pore within the plasma membrane. Further, CD59 is a low-affinity ligand of human CD2, causes T cell costimulation, and is involved in lymphocyte signal transduction. CD59 is a potent inhibitor of the complement membrane attack complex, whereby it binds complement C8 and/or C9 during the assembly of this complex, thereby inhibiting the incorporation of multiple copies of C9 into the complex, which is necessary for osmolytic pore formation. Mutations in this gene cause CD59 deficiency, a disease involving hemolytic anemia, thrombosis, and cerebral infarction. Multiple alternatively spliced transcript variants of CD59 have been identified.Specifications
CD59 | |
Polyclonal | |
Unconjugated | |
CD59 | |
16.3A5, 1F5, 1F5 antigen, CD59, CD59 glycoprotein, EJ16, EJ30, EL32, G344, HRF 20, HRF20, MAC inhibitory protein, MAC IP, MACIF, MEM43, MEM43 antigen, MIC11, MIN1, MIN2, MIN3, MIRL, MSK21, p18 20, Protectin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
966 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P13987 | |
CD59 | |
CD59 Fusion Protein Ag1173 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction