missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD31 (PECAM-1) Chimeric Recombinant Rabbit Monoclonal Antibody (BAG-85D10), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA548260
This item is not returnable.
View return policy
Description
Specificity: This antibody recognises CD31 (PECAM1) which is a member of the immunoglobulin superfamily and is expressed at various circulatng cells, such as neutrophils, monocytes, platelets as well as on endothelial cells. CD31 is a cell adhesion molecule and is involved in the leukocyte transendothelial migration during inflammation.
CD31 (platelet endothelial cell adhesion molecule-1, PECAM-1) is an inhibitory coreceptor involved in regulation of T cell and B cell signaling by a dual immunoreceptor tyrosine-based inhibitory motif (ITIM) that upon associated kinases-mediated phosphorylation provide docking sites for protein-tyrosine phosphatases. CD31 is expressed ubiquitously within the vascular compartment and is located mainly at junctions between adjacent cells. N-terminal Ig-like domain of CD31 is responsible for its homophilic binding, which plays an important role in cell-cell interactions. CD31 is a multifunctional molecule with diverse roles in modulation of integrin-mediated cell adhesion, transendothelial migration, angiogenesis, apoptosis, negative regulation of immunoreceptor signaling, autoimmunity, macrophage phagocytosis, IgE-mediated anaphylaxis and thrombosis. It is one of key regulatory molecules in vascular system.Specifications
CD31 (PECAM-1) Chimeric | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
P16284 | |
Pecam1 | |
This antibody was raised by immunising mice with human peripheral blood lymphocytes from a patient with high numbers of gamma-delta T cells. | |
200 μg | |
Primary | |
Human | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry | |
BAG-85D10 | |
Unconjugated | |
Pecam1 | |
C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule | |
Rabbit | |
Affinity chromatography | |
RUO | |
5175 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG κ |