Learn More
CD226 (DNAM-1) Monoclonal Antibody (C1), Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen MA543904
 View more versions of this product
Product Code. 17684356
  • 1632.00 NOK / 20µL
Explore more special offers
Description
Description
DNAM 1 is a receptor expressed by peripheral blood T lymphocytes that is involved in intercellular adhesion, lymphocyte signaling, cytotoxicity and lymphokine secretion mediated by cytotoxic T lymphocytes. DNAM 1 is broadly expressed on T cells, NK cells, platelets, monocytes and a subset of B cells. DNAM-1 is also expressed by a subset of CD3 positive thymocytes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD226 (DNAM-1)
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300
Q15762
Cd226
Glu19-Ala254 (Accession Q15762), with N-terminal His Tag
20 μL
Primary
Human
Antibody
IgG1 κ
Western Blot
C1
Unconjugated
Cd226
adhesion glycoprotein; BC051526; Cd226; CD226 antigen; CD226 molecule; DNAM 1; DNAM1; DNAM-1; DNAX accessory molecule 1; DNAX accessory molecule-1; platelet and T cell activation antigen 1; platelet and T-cell activation antigen 1; Pta1; T lineage-specific activation antigen 1 antigen; TLiSA1
Mouse
Protein A/G
RUO
10666
-20°C