missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Chimeric Recombinant Rabbit Monoclonal Antibody (4G7), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA547845
3745.00 NOK valid until 2025-03-16
Use promo code "24070" to get your promotional price.
This item is not returnable.
View return policy
Description
Specificity: Recognises human CD19.
CD19 is a member of the immunoglobulin superfamily and has two Ig like domains. The CD19 molecule is expressed on 100% of the peripheral B cells as defined by expression of kappa or lambda light chains. CD19 appears to be expressed on myeloid leukemia cells, particularly those of monocytic lineage. Leukemia phenotype studies have demonstrated that the earliest and broadest B cell restricted antigen is the CD19 antigen. The receptor for CD19 is an important functional regulator of normal and malignant B cell proliferation, and is expressed in all B cell precursor leukemias. Lymphocytes proliferate and differentiate in response to various concentrations of different antigens. The ability of the B cell to respond in a specific, yet sensitive manner to the various antigens is achieved with the use of low-affinity antigen receptors. CD19 is a cell surface molecule which assembles with the antigen receptor of B lymphocytes in order to decrease the threshold for antigen receptor-dependent stimulation. Besides being a signal-amplifying coreceptor for the B cell receptor (BCR), CD19 can also signal independently of BCR co-ligation and is a central regulatory component upon which multiple signaling pathways converge. Mutation of the CD19 gene results in hypogammaglobulinemia, whereas CD19 overexpression causes B cell hyperactivity.
Specifications
CD19 Chimeric | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
P15391 | |
Cd19 | |
Human CCL (chronic lymphocytic leukemia) cells. | |
200 μg | |
Primary | |
Human | |
Liquid |
Flow Cytometry, Immunocytochemistry | |
4G7 | |
Unconjugated | |
Cd19 | |
AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12 | |
Rabbit | |
Affinity chromatography | |
RUO | |
930 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD19 Chimeric Recombinant Rabbit Monoclonal Antibody (4G7), Invitrogen™ > 200 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction