Learn More
CD161f Rat anti-Mouse, BV605, Clone: C8, BD OptiBuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 743513
4483.30 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
CD161f Monoclonal antibody specifically detects CD161f in Mouse samples. It is validated for Flow Cytometry.
Specifications
CD161f | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
C8 | |
Brilliant Violet 605 | |
50 ÎĽg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 Îş |
Product Content Correction
Product Title
CD161f Rat anti-Mouse, BV605, Clone: C8, BD OptiBuild™ > Ms CD161f BV605 C8 50 μg