Learn More
CCS Viscosity Standard, ASTM D 5293 and SAE Specification J310, Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified CCS viscosity standards are for the calibration and verification of analytical equipment used in Cold Cranking Simulator (CCS) testing to ASTM D5293 and SAE Specification J300.
Brand:  Paragon Scientific CL16
Product Code. 16060010
  • 1040.00 NOK / 500mL
Explore more special offers
Description
Description
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Full Traceability to National Standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Manufactured in the United Kingdom
  • 2 year shelf life
Specifications
Specifications
Glass Amber Bottle
500 mL
Yes
Liquid
Product Title
CCS Viscosity Standard, ASTM D 5293 and SAE Specification J310, Paragon Scientific™ > Quantity: 500mL
Nominal values quoted. Certified Values will vary from batch to batch.