missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL8/MCP-2 Mouse anti-Human, Clone: CCL8/3312, Novus Biologicals™
Mouse Monoclonal Antibody
2355.00 NOK - 4680.00 NOK
Specifications
Antigen | CCL8/MCP-2 |
---|---|
Clone | CCL8/3312 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Immunohistochemistry (Paraffin), Peptide Array |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18300576
|
Bio-Techne
NBP3-13769-20UG |
20 μg |
2355.00 NOK
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18329681
|
Bio-Techne
NBP3-13769-100UG |
100 μg |
4680.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL8/MCP-2 Monoclonal antibody specifically detects CCL8/MCP-2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
CCL8/MCP-2 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research, Signal Transduction | |
10 mM PBS with 0.05% BSA | |
C-C motif chemokine 8, chemokine (C-C motif) ligand 8, HC14, MCP2, MCP-2, member 8 (monocyte chemotactic protein 2), monocyte chemoattractant protein 2, SCYA10, small-inducible cytokine A8 | |
Human recombinant CCL8/MCP-2 protein fragment (around aa24-98) (exact sequence is proprietary) (Uniprot: P80075) | |
Primary | |
Store at 4°C. |
CCL8/3312 | |
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P80075 | |
6355 | |
IgG2b κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title