missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals CCL11/Eotaxin Antibody (1C8-2C1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00006356-M01
This item is not returnable.
View return policy
Description
CCL11/Eotaxin Monoclonal antibody specifically detects CCL11/Eotaxin in Human samples. It is validated for Western Blot, ELISA
Specifications
CCL11/Eotaxin | |
Monoclonal | |
Unconjugated | |
In 1x PBS, pH 7.4 | |
C-C motif chemokine 11, chemokine (C-C motif) ligand 11, Eosinophil chemotactic protein, eotaxin, eotaxin-1, MGC22554, SCYA11Small-inducible cytokine A11, small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin) | |
CCL11 (AAH17850, 24 a.a. ~ 97 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GPASVPTTCCFNLANRKIPLQRLESYRRITSGKCPQKAVIFKTKLAKDICADPKKKWVQDSMKYLDQKSPTPKP | |
0.1 mg | |
Asthma, Cancer, Cytokine Research, Immunology | |
6356 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG1 Îş |
Western Blot, ELISA | |
1C8-2C1 | |
Western Blot, ELISA | |
AAH17850 | |
Mouse | |
IgG purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals CCL11/Eotaxin Antibody (1C8-2C1), Novus Biologicals™ > 0.1 mg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction