Learn More
Motic™ CCIS™ Plan Achromatic Strain-free Objectives
Plan Achromatic strain-free objectives for use with TEC POL Microscopes.
Brand:  Motic™ 1101001707221
 View more versions of this product
Product Code. 17128628
  • 4220.00 NOK / Each
Estimated Shipment: 17-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with Motic™ Panthera TEC POL Microscopes
  • UC PL P Plan Achromatic strain-free objectives
Specifications
Specifications
Microscope Objective
Plan Achromatic
0.65
Motic™ TEC POL Microscopes
40X
0.6 mm