Learn More
Brady™ Caution Labels
Brand:  Brady™ 225251
 View more versions of this product
Product Code. 11776825
  • 207.00 NOK / 1 pack
Only null left
Estimated Shipment: 26-06-2025
Specifications
10 Pk.
TAG-NE PAS METTRE EN M.-50X110MM
Polyester
Lockout Tagout
110 mm
2 in.
-40°C to +100°C
1.97 in.
7 mm
Tag
White on Red
Ne pas mettre en marche Equipe d’entretien Cette etiquette ne peut etre enlevee que par:
4.33 in.
50 mm
0.249 mm
-40°F to +212°F
50 mm
Lockout Tagout Tags
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Brady™ Caution Labels > Caution Labels; Message: Do not Start this condemnation; Dimensions (L x W): 50 mm x 110 mm