Specifications
10 Pk. | |
TAG-NE PAS COUPER-50X110MM | |
Polyester | |
Lockout Tagout | |
110 mm | |
4.3 in. | |
-40°C to +100°C | |
1.97 in. | |
7 mm |
Tag | |
White on Red | |
Ne pas couper l’electricite | |
4.33 in. | |
110 mm | |
0.249 mm | |
-40°F to +212°F | |
50 mm | |
Lockout Tagout Tags |
Product Content Correction
Product Title
Brady™ Caution Labels > Caution Labels; Message: Do not cut Electricity; Dimensions (L x W): 110 mm x 50 mm