Learn More
Brady™ Caution Labels
Brand:  Brady™ 225238
 View more versions of this product
Product Code. 11746815
  • 207.00 NOK / 1 pack
Only null left
Estimated Shipment: 27-06-2025
Specifications
10 Pk.
TAG-VIERGE FR-50X110MM
Polyester
Lockout Tagout
110 mm
4.3 in.
-40°C to +100°C
1.97 in.
7 mm
Tag
Red on White
Condemnation
4.33 in.
110 mm
0.249 mm
-40°F to +212°F
50 mm
Lockout Tagout Tags
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Brady™ Caution Labels > Caution Labels; Message: Blank; Dimensions (L x W): 110 mm x 50 mm