Specifications
10 Pk. | |
TAG-DEFENSE D'OUV R.-50X110MM | |
Polyester | |
Lockout Tagout | |
110 mm | |
2 in. | |
-40°C to +100°C | |
1.97 in. | |
7 mm |
Tag | |
White on Red | |
Defense d’ouvrir la vanne | |
4.33 in. | |
50 mm | |
0.249 mm | |
-40°F to +212°F | |
50 mm | |
Lockout Tagout Tags |
Product Content Correction
Product Title
Brady™ Caution Labels > Caution Labels; Message: Defence to open the valve; Dimensions (L x W): 50 mm x 110 mm