Learn More
Brady™ Caution Labels
Brand:  Brady™ 230776
 View more versions of this product
Product Code. 11706845
  • 285.00 NOK / 1 pack
Only null left
Estimated Shipment: 25-06-2025
Specifications
10 Pk.
TAG-F DANGER NE PAS METTRE-75X160MM
Polyester
Lockout Tagout
160 mm
-40°C to +100°C
2.95 in.
7 mm
Tag
Black, Red on White
DANGER NE PAS METTRE EN ROUTE SANS L’AUTORISATION
6.3 in.
0.249 mm
-40°F to +212°F
75 mm
Lockout Tagout Tags
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Brady™ Caution Labels > Caution Labels; Message: Risk associated with product and machinery; Dimensions (L x W):