Learn More
Swann-Morton™ Cast Metal Fixed handle
Brand: Swann-Morton™ 4302
This item is not returnable.
View return policy
Specifications
Heavy Duty Cast Metal | |
SM00 Fixed Handle | |
123 mm | |
Choice of 4 Heavy-Duty Blade: SM01, SM02, SM03 and SM04 |
Product Content Correction
Product Title
Swann-Morton™ Cast Metal Fixed handle > Length: 123mm