missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspr2 Rabbit anti-Human, Mouse, Rat, Clone: 2H2I5, Novus Biologicals™
Rabbit Monoclonal Antibody
1815.00 NOK - 4580.00 NOK
Specifications
Antigen | Caspr2 |
---|---|
Clone | 2H2I5 |
Dilution | Western Blot 1:500 - 1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18349165
|
Bio-Techne
NBP3-15723-20UL |
20 μg |
1815.00 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18329664
|
Bio-Techne
NBP3-15723-100UL |
100 μg |
4580.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caspr2 Monoclonal antibody specifically detects Caspr2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Caspr2 | |
Western Blot 1:500 - 1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
AUTS15, CASPR2DKFZp781D1846, CDFE, Cell recognition molecule Caspr2, contactin associated protein-like 2, contactin-associated protein 2, contactin-associated protein-like 2, homolog of Drosophila neurexin IV, KIAA0868, NRXN4Caspr2, PTHSL1 | |
A synthetic peptide corresponding to a sequence within amino acids 1232-1331 of human Caspr2 (Q9UHC6). HLDHLDSASADFPYNPGQGQAIRNGVNRNSAIIgGVIAVVIFTILCTLVFLIRYMFRHKGTYHTNEAKGAESAESADAAIMNNDPNFTETIDESKKEWLI | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
2H2I5 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
26047 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title