missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 19 publications
Brand: Novus Biologicals NB100-56118SS
This item is not returnable.
View return policy
Description
Caspase-9 Polyclonal specifically detects Caspase-9 in Human, Mouse, Rat, Canine, Gerbil samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).
Specifications
Caspase-9 | |
Polyclonal | |
Western Blot reported in scientific literature (PMID 24184144), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 22223358), Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen Assay Dependent, Flow (Intracellular) | |
P55211 | |
CASP9 | |
Human Caspase 9 recombinant protein catalytic subunits (NP_001220). | |
0.025 mL | |
Alzheimers Research, Apoptosis, Cancer, Caspases, Cell Biology, Neurodegeneration | |
842 | |
Human, Mouse, Rat, Canine, Gerbil | |
Antisera |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Whole antisera with 0.05% Sodium Azide | |
APAF-3APAF3, apoptotic protease activating factor 3, Apoptotic protease Mch-6, Apoptotic protease-activating factor 3, CASP-9, caspase 9, apoptosis-related cysteine peptidase, caspase-9, EC 3.4.22.62, ICE-LAP6CASPASE-9c, ICE-like apoptotic protease 6, MCH6apoptosis-related cysteine protease | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
This antibody detects both the pro- and active-Caspase 9 forms. In western blots, the proform of caspase-9 is detected at ∼50 kDa, the large subunit at ∼35 kDa, and the small subunit at ∼15 kDa. Intermediate caspase-9 cleavage forms may also be seen at ∼21 kDa. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Caspase-9 Antibody, Novus Biologicals™ > 0.025mL; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction