Learn More
Thermo Scientific™ Barnstead™ Water System Cartridges
Maintain your Thermo Scientific™ Barnstead™ point of use water purification system with these replacement cartridges.
Brand: Thermo Scientific™ 09.1020
6211.16 NOK valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Description
Maintain your Thermo Scientific™ Barnstead™ point of use water purification systems with replacement Thermo Scientific™ Barnstead™ Cartridges.
Choose the appropriate part number for your system. These cartridges are for use with the following Barnstead point of use water purification systems:- MicroPure™
- GenPure™
- GenPure™ Pro
- GenPure™ xCAD
- Smart2Pure™
- LabTower™ EDI
- Pacific™ TII
Specifications
Ultrapure polisher cartridge | |
Smart2Pure | |
Cartridge, Ultrapure Polisher |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59% -
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30